Anti-PI3K p110gamma Rabbit Polyclonal Antibody [clone: L89]
Catalog # BWRLBS9175
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PI3K p110 gamma
- Clonality:Polyclonal
- Clone:L89
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human PI3K p110gamma.
PI 3-Kinase p110? (L89) pAb detects endogenous levels of PI 3-Kinase p110? protein.
Type: Primary
Antigen: PI3K p110gamma
Clonality: Polyclonal
Clone: L89
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human