Anti-ZNF262 Rabbit Polyclonal Antibody [clone: H837]
BWRLBS9170
- Antibody type:Primary
- Antigen symbol:ZNF262
- Clonality:Polyclonal
- Clone:H837
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 804-856 of Human ZNF262.
ZNF262 (H837) pAb detects endogenous levels of ZNF262 protein.
Type: Primary
Antigen: ZNF262
Clonality: Polyclonal
Clone: H837
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse