Specifications
- Antibody type:Primary
- Antigen symbol:WIP
- Clonality:Polyclonal
- Clone:Y455
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 431-480 of Human WIP.
WIP (Y455) pAb detects endogenous levels of WIP protein.
Type: Primary
Antigen: WIP
Clonality: Polyclonal
Clone: Y455
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat