Anti-GPR38 Rabbit Polyclonal Antibody
Catalog # BWRLBS5749
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GPR38
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 367-412 of Human GPR38.
GPR38 pAb detects endogenous levels of GPR38 protein.
Type: Primary
Antigen: GPR38
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat