Anti-p-SRF Rabbit Polyclonal Antibody [clone: S103]
Catalog # BWRLBS4177
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-SRF
- Clonality:Polyclonal
- Clone:S103
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human SRF around the phosphorylation site of Serine 103.
p-SRF (S103) pAb detects endogenous levels of SRF protein when phosphorylated at Ser103.
Type: Primary
Antigen: p-SRF
Clonality: Polyclonal
Clone: S103
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat