Order Entry
Ireland
ContactUsLinkComponent
Anti-EEF1A1 Rabbit Polyclonal Antibody
Anti-EEF1A1 Rabbit Polyclonal Antibody
Catalog # BWRLBS6077
Anti-EEF1A1 Rabbit Polyclonal Antibody
Catalog # BWRLBS6077

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EEF1A1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human EEF1A1.

EEF1A1 pAb detects endogenous levels of EEF1A1 protein.

Type: Primary
Antigen: EEF1A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat