Anti-DHRS7 Rabbit Polyclonal Antibody
Catalog # BWRLBS60139
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:DHRS7
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human DHRS7.
DHRS7 pAb detects endogenous levels of DHRS7 protein.
Type: Primary
Antigen: DHRS7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat