Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ABCD3 Rabbit Polyclonal Antibody
  BWRLBS60111
Anti-ABCD3 Rabbit Polyclonal Antibody
  BWRLBS60111

 

  • Antibody type:
    Primary
  • Antigen symbol:
    ABCD3
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

A synthetic peptide corresponding to residues in Human ABCD3.

ABCD3 pAb detects endogenous levels of ABCD3 protein.

Type: Primary
Antigen: ABCD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat