Anti-p-Raf-1 Rabbit Polyclonal Antibody [clone: S338]
Catalog # BWRLBS4732
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Raf-1
- Clonality:Polyclonal
- Clone:S338
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Raf-1 around the phosphorylation site of Serine 338.
p-Raf-1 (S338) pAb detects endogenous levels of Raf1 protein only when phosphorylated at Ser338
Type: Primary
Antigen: p-Raf-1
Clonality: Polyclonal
Clone: S338
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat