Anti-CSF2 Rabbit Polyclonal Antibody
Catalog # BWRLBS60003
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CSF2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human CSF2.
CSF2 pAb detects endogenous levels of CSF2 protein.
Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat