Anti-p-c-Src Rabbit Polyclonal Antibody [clone: S75]
BWRLBS4674
- Antibody type:Primary
- Antigen symbol:p-c-Src
- Clonality:Polyclonal
- Clone:S75
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human c-Src around the phosphorylation site of Serine 75.
p-Src (S75) pAb detects endogenous levels of Src protein when phosphorylated at Ser75.
Type: Primary
Antigen: p-c-Src
Clonality: Polyclonal
Clone: S75
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat