Anti-p-CHOP Rabbit Polyclonal Antibody [clone: S30]
Catalog # BWRLBS4671
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CHOP
- Clonality:Polyclonal
- Clone:S30
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CHOP around the phosphorylation site of Serine 30.
p-CHOP (S30) pAb detects endogenous levels of CHOP protein only when phosphorylated at Ser30.
Type: Primary
Antigen: p-CHOP
Clonality: Polyclonal
Clone: S30
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat