Anti-ASK1 Rabbit Polyclonal Antibody
Catalog # BWRLBS6445
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ASK1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human ASK1.
ASK1 pAb detects endogenous levels of ASK1 protein.
Type: Primary
Antigen: ASK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat