Anti-Aldolase A Rabbit Polyclonal Antibody [clone: T9]
Catalog # BWRLBS1912
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Aldolase A
- Clonality:Polyclonal
- Clone:T9
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human Aldolase A.
Aldolase A (T9) pAb detects endogenous levels of Aldolase A protein.
Type: Primary
Antigen: Aldolase A
Clonality: Polyclonal
Clone: T9
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat