Order Entry
Ireland
ContactUsLinkComponent
Anti-Dyskerin Rabbit Polyclonal Antibody [clone: K203]
Anti-Dyskerin Rabbit Polyclonal Antibody [clone: K203]
  BWRLBS1887
Anti-Dyskerin Rabbit Polyclonal Antibody [clone: K203]
  BWRLBS1887

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Dyskerin
  • Clonality:
    Polyclonal
  • Clone:
    K203
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 171-220 of Human Dyskerin.

Dyskerin (K203) pAb detects endogenous levels of Dyskerin protein.

Type: Primary
Antigen: Dyskerin
Clonality: Polyclonal
Clone: K203
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat