Anti-Casein Kinase IIalpha Rabbit Polyclonal Antibody [clone: T251]
Catalog # BWRLBS1859
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Casein Kinase II alpha
- Clonality:Polyclonal
- Clone:T251
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 220-270 of Human Casein Kinase IIalpha.
Casein Kinase II? (T251) pAb detects endogenous levels of Casein kinase II subunit alpha protein.
Type: Primary
Antigen: Casein Kinase IIalpha
Clonality: Polyclonal
Clone: T251
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat