Anti-COP zeta1 Rabbit Polyclonal Antibody [clone: I48]
BWRLBS3287
- Antibody type:Primary
- Antigen symbol:COP zeta1
- Clonality:Polyclonal
- Clone:I48
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human COP zeta1.
COP ?1 (I48) pAb detects endogenous levels of COP ?1 protein.
Type: Primary
Antigen: COP zeta1
Clonality: Polyclonal
Clone: I48
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat