Anti-Nur77/NR4A1 Rabbit Polyclonal Antibody [clone: G347]
Catalog # BWRLBS3260
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Nur77/NR4A1
- Clonality:Polyclonal
- Clone:G347
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 320-365 of Human Nur77/NR4A1.
Nur77/NR4A1 (G347) pAb detects endogenous levels of Nur77/NR4A1 protein.
Type: Primary
Antigen: Nur77/NR4A1
Clonality: Polyclonal
Clone: G347
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat