Anti-HSL Rabbit Polyclonal Antibody [clone: A546]
Catalog # BWRLBS2742
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HSL
- Clonality:Polyclonal
- Clone:A546
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 520-570 of Human HSL.
HSL (A546) pAb detects endogenous levels of HSL protein.
Type: Primary
Antigen: HSL
Clonality: Polyclonal
Clone: A546
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat