Anti-MARK1/2/3/4 Rabbit Polyclonal Antibody [clone: N211]
BWRLBS3001
- Antibody type:Primary
- Antigen symbol:MARK1/2/3/4
- Clonality:Polyclonal
- Clone:N211
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 180-230 of Human MARK1.
MARK1/2/3/4 (N211) pAb detects endogenous levels of MARK1/2/3/4 protein.
Type: Primary
Antigen: MARK1/2/3/4
Clonality: Polyclonal
Clone: N211
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat