Anti-AMPKgamma1/2/3 Rabbit Polyclonal Antibody [clone: K78]
Catalog # BWRLBS2480
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AMPK gamma1/2/3
- Clonality:Polyclonal
- Clone:K78
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human AMPKgamma1.
AMPK?1/2/3 (K78) pAb detects endogenous levels of total AMPK? protein.
Type: Primary
Antigen: AMPKgamma1/2/3
Clonality: Polyclonal
Clone: K78
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat