Anti-TTF-1 Rabbit Polyclonal Antibody [clone: V27]
BWRLBS2947
- Antibody type:Primary
- Antigen symbol:TTF-1
- Clonality:Polyclonal
- Clone:V27
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminal of Human TTF-1.
TTF-1 (V27) pAb detects endogenous levels of TTF-1 protein.
Type: Primary
Antigen: TTF-1
Clonality: Polyclonal
Clone: V27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat