Anti-MSK1 Rabbit Polyclonal Antibody [clone: M355]
BWRLBS1553
- Antibody type:Primary
- Antigen symbol:MSK1
- Clonality:Polyclonal
- Clone:M355
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 331-380 of Human MSK1.
MSK1 (M355) pAb detects endogenous levels of MSK1 protein.
Type: Primary
Antigen: MSK1
Clonality: Polyclonal
Clone: M355
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat