Anti-Kv3.4 Rabbit Polyclonal Antibody [clone: R11]
Catalog # BWRLBS3728
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Kv3.4
- Clonality:Polyclonal
- Clone:R11
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human KV3.4.
Kv3.4 (R11) pAb detects endogenous levels of Kv3.4 protein.
Type: Primary
Antigen: Kv3.4
Clonality: Polyclonal
Clone: R11
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse