Anti-SURF-1 Rabbit Polyclonal Antibody [clone: F181]
BWRLBS2068
- Antibody type:Primary
- Antigen name:Surfeit 1
- Antigen symbol:SURF1
- Clonality:Polyclonal
- Clone:F181
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 150-200 of Human SURF-1.
SURF-1 (F181) pAb detects endogenous levels of SURF-1 protein.
Type: Primary
Antigen: SURF-1
Clonality: Polyclonal
Clone: F181
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat