Anti-Sp3/4 Rabbit Polyclonal Antibody [clone: G659]
Catalog # BWRLBS2030
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Sp3/4
- Clonality:Polyclonal
- Clone:G659
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 630-680 of Human Sp3/4.
Sp3/4 (G659) pAb detects endogenous levels of Sp3/4 protein.
Type: Primary
Antigen: Sp3/4
Clonality: Polyclonal
Clone: G659
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat