Anti-p-Adducin alpha/beta Rabbit Polyclonal Antibody [clone: S726/713]
Catalog # BWRLBS4005
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Adducin alpha/beta
- Clonality:Polyclonal
- Clone:S726/713
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Adducin alpha around the phosphorylation site of Serine 726.
p-Adducin ?/? (S726/713) pAb detects endogenous levels of Adducin ? protein only when phosphorylated at Ser726 and Adducin ? protein only when phosphorylated at Ser713.
Type: Primary
Antigen: p-Adducin alpha/beta
Clonality: Polyclonal
Clone: S726/713
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat