Order Entry
Ireland
ContactUsLinkComponent
Anti-p-14-3-3 zeta Rabbit Polyclonal Antibody [clone: S58]
Anti-p-14-3-3 zeta Rabbit Polyclonal Antibody [clone: S58]
Catalog # BWRLBS4001
Anti-p-14-3-3 zeta Rabbit Polyclonal Antibody [clone: S58]
Catalog # BWRLBS4001

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-14-3-3 zeta
  • Clonality:
    Polyclonal
  • Clone:
    S58
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Histone 14-3-3 zeta around the phosphorylation site of Serine 58.

p-14-3-3 ? (S58) pAb detects endogenous levels of 14-3-3 protein zeta/delta only when phosphorylated at Ser58.

Type: Primary
Antigen: p-14-3-3 zeta
Clonality: Polyclonal
Clone: S58
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat