Anti-ELL Rabbit Polyclonal Antibody [clone: D125]
BWRLBS2020
- Antibody type:Primary
- Antigen symbol:ELL
- Clonality:Polyclonal
- Clone:D125
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 100-150 of Human ELL.
ELL (D125) pAb detects endogenous levels of ELL protein.
Type: Primary
Antigen: ELL
Clonality: Polyclonal
Clone: D125
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat