Order Entry
Ireland
ContactUsLinkComponent
 
Anti-MEK-6 Rabbit Polyclonal Antibody [clone: Q3]
  BWRLBS2013
Anti-MEK-6 Rabbit Polyclonal Antibody [clone: Q3]
  BWRLBS2013

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK-6
  • Clonality:
    Polyclonal
  • Clone:
    Q3
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids N-terminus of Human MEK-6.

MEK-6 (Q3) pAb detects endogenous levels of MEK-6 protein.

Type: Primary
Antigen: MEK-6
Clonality: Polyclonal
Clone: Q3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat