Anti-MOX-2 Rabbit Polyclonal Antibody [clone: N184]
BWRLBS2006
- Antibody type:Primary
- Antigen symbol:MOX-2
- Clonality:Polyclonal
- Clone:N184
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 150-200 of Human MOX-2.
MOX-2 (N184) pAb detects endogenous levels of MOX-2 protein.
Type: Primary
Antigen: MOX-2
Clonality: Polyclonal
Clone: N184
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat