Anti-HXK I Rabbit Polyclonal Antibody [clone: F67]
Catalog # BWRLBS1947
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HXK I
- Clonality:Polyclonal
- Clone:F67
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 30-80 of Human HXK I.
HXK I (F67) pAb detects endogenous levels of HXK I protein.
Type: Primary
Antigen: HXK I
Clonality: Polyclonal
Clone: F67
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat