Order Entry
Ireland
ContactUsLinkComponent
Anti-UBF-1 Rabbit Polyclonal Antibody [clone: E536]
Anti-UBF-1 Rabbit Polyclonal Antibody [clone: E536]
Catalog # BWRLBS1939
Anti-UBF-1 Rabbit Polyclonal Antibody [clone: E536]
Catalog # BWRLBS1939

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    UBF-1
  • Clonality:
    Polyclonal
  • Clone:
    E536
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 500-550 of Human UBF-1.

UBF-1 (E536) pAb detects endogenous levels of UBF-1 protein.

Type: Primary
Antigen: UBF-1
Clonality: Polyclonal
Clone: E536
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat