Anti-GluR-5 Rabbit Polyclonal Antibody [clone: L12]
Catalog # BWRLBS1145
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GluR-5
- Clonality:Polyclonal
- Clone:L12
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids N-terminus of Human GluR-5.
GluR5(L12) pAb detects endogenous levels of GluR5 protein.
Type: Primary
Antigen: GluR-5
Clonality: Polyclonal
Clone: L12
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat