Order Entry
Ireland
ContactUsLinkComponent
FSC 22® embedding compound, Surgipath®
  LEIC3801480
 :  
FSC 22® embedding compound, Surgipath®
  LEIC3801480
 :  
  • Pk:
    1
  • Description:
    FSC 22, frozen, 9×118 ml
This FSC 22® is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning.
  • Provides consistent sectioning with minimal curling of sections
  • Working temperature −20 °C
  • Available in clear for better visualisation of small specimens