Order Entry
Ireland
ContactUsLinkComponent
Guanidinium hydrochloride ≥99%, white crystalline powder, Ultrapure
Irritant
Guanidinium hydrochloride ≥99%, white crystalline powder, Ultrapure
Supplier:  MP Biomedicals
Certificates
undefined
Irritant
Guanidinium hydrochloride ≥99%, white crystalline powder, Ultrapure
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • pH (6.0 M aq soln):
    4-6
  • OD260 (6.0M aq soln):
    ≤0.03
  • Loss on Drying:
    ≤1.0%
  • Solubility (6.0 M aq soln):
    Clear, Colorless Solution

Specification Test Results

Related Information

Frequently Bought Together