Vitamin K1 (phytomenadione, phylloquinone), thick amber liquid
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063
Boiling Pt:∼140 °C (0.001 m) (lit.)
Melting Pt:–20 °C (lit.)
Density:0.984 (lit.)
Molecular Weight:450.6958 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test A:Positive
- Identity Test B:Positive
- Reaction to Litmus (5% in alcohol):Neutral
- Refractive Index @ 25°C:1.523-1.526
- Menadione:Not Detected
- Z Isomer Content:≤21.0%
- Assay (Total E and Z Isomers):≥97.0%
Specification Test Results
Frequently Bought Together