Order Entry
Ireland
ContactUsLinkComponent
Anti-ALDOA Rabbit Polyclonal Antibody
Anti-ALDOA Rabbit Polyclonal Antibody
Catalog # ABNOPAB30682
Supplier:  Abnova
Anti-ALDOA Rabbit Polyclonal Antibody
Catalog # ABNOPAB30682
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ALDOA Recombinant
  • Antigen symbol:
    ALDOA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    226
  • Storage buffer:
    In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
  • Sequence:
    IVAPGKGILAADESTGSIAKRLQSIGTENTEENRRFYRQLLLTADDRVNPCIGGVILFHETLYQKADDGRPFPQVIKSKGGVVGIKVDKGVVPLAGTNGETTTQGL
  • Storage temperature:
    Store at 4° C. For long term storage store at -20° C.
  • Immunogen:
    Recombinant ALDOA.
  • Purification:
    Antigen affinity purification
  • Size:
    100 µL
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against partial recombinant human ALDOA.

Type: Primary
Antigen: ALDOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human