Pierce™ Western Blot Signal Enhancer
Catalog # PIER21050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 ml
- Description:Western Blot Signal Enhancer
Specifications
About this item
Western blot signal enhancer is a membrane treatment that has been shown to increase intensity of a target protein band on a Western blot and to enable detection of a target analyte at a Level at which it could not be detected without enhancement.
- Protein targets can be detected at lower levels
- Enhances antibody binding to immobilised antigens
- Works with any substrate, enhancing both chemiluminescence and colorimetric detection
- Works with any membrane
- Ready-to-use and can be stored at room temperature