Latrunculin A
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one
CAS Number:76343-93-6
:
Formula:C₂₂H₃₁NO₅S
Molecular Weight:421.552 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together