Order Entry
Ireland
ContactUsLinkComponent
Miracloth, Calbiochem®
Miracloth, Calbiochem®
Catalog # 475855-1
Miracloth, Calbiochem®
Catalog # 475855-1
Specifications
  • Pore size:
    22,00 - 25,00 µm
  • Description:
    Miracloth
  • Environmentally Preferable:
  • Pk:
    1 Roll
Specifications
About this item
Miracloth is an autoclavable, quick filtration material for gelatinous homogenates that is composed of rayon-polyester with an acrylic binder. Ideal for filtration of cleared neutralised lysates during medium- and large-scale plasmid purification to prevent contamination with loosely adherent pellet material and is useful for protoplast isolation.