Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
beta-1,4-Glucosidase
beta-1,4-Glucosidase
beta-1,4-Glucosidase
Catalog # AGRIAS153087
Supplier:
Agrisera
CAS Number:
Expand image
beta-1,4-Glucosidase
Catalog # AGRIAS153087
Supplier:
Agrisera
CAS Number:
Specifications
Pk:
50 µl
Storage conditions:
Store lyophilized/reconstituted at –20 °C
Form:
Lyophilized
Enzyme name:
beta-1,4-glucosidase
Specifications
About this item
Product Details
Documentation
Product Details
Documentation