Order Entry
Ireland
ContactUsLinkComponent
Human IL8 (from E. coli)
Human IL8 (from E. coli)
Supplier:  Biorbyt
Certificates
Human IL8 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized Interleukin-8 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CXCL8 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    Specific Activity of IL8 in chemotaxis of donor PBL neutrophils, threshold concentration corresponding to 10-100 ng/ml.
  • Protein synonyms:
    310C
  • Protein/peptide name:
    IL8
  • Purity:
    Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin-8 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Ala-Lys-Glu-Leu.
  • Formulation:
    The IL-8 was lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together