Order Entry
Ireland
ContactUsLinkComponent
 
Human CT 1 (from E. coli)
 :  Biorbyt
 
Human CT 1 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized Cardiophin remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    CTF1
  • Protein/peptide name:
    CT 1
  • Purity:
    Purity of CTF1 Human Recombinant is greater than 90% as determined by SDS-PAGE.
  • Solubility:
    Add 0.2 uml of 0.1M Acetate buffer pH-4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of thi
  • Formulation:
    Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH-4.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together