SARS matrix
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month at 4 °C.
- Protein/peptide name:SARS Matrix
- Purity:SARS Matrix Protein is >95% pure as determined by 10% PAGE (coomassie staining). SARS Matrix protein was purified by proprietary chromatographic technique.
- Formulation:50mM Tris-HCl, 60mM NaCl, and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together