Order Entry
Ireland
ContactUsLinkComponent
HCV NS3 genotype-2c
HCV NS3 genotype-2c
Supplier:  Biorbyt
Certificates
HCV NS3 genotype-2c
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HCV NS3 Genotype-2c
  • Purity:
    HCV NS3 Genotype-2c protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV NS3 Genotype-2c protein was purified by proprietary chromatographic technique.
  • Formulation:
    1.5M urea, 25mM Tris-HCl pH-8, 0.2% Triton-X 50% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together