HCV genotype-1
Marketsource
: Biorbyt
Documents
orb81554
Datasheet
- Catalog No:
- BIRBORB81554-100
- BIRBORB81554-500
- Protein/peptide name:
- HCV Genotype-1
- HCV Genotype-1
- Purity:
- HCV Core Genotype-1 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-1 protein was purified by proprietary chromatographic technique.
- HCV Core Genotype-1 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-1 protein was purified by proprietary chromatographic technique.
- Formulation:
- 50mM Tris. pH-8 5mM EDTA.
- 50mM Tris. pH-8 5mM EDTA.
- Storage conditions:
- Five years frozen. One month in solution at room temperature.
- Five years frozen. One month in solution at room temperature.
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Product Family Options
- Pk
- Marketsource
Specifications
Cat. No.BIRBORB81554-100Protein/peptide nameHCV Genotype-1PurityHCV Core Genotype-1 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-1 protein was purified by proprietary chromatographic technique.Formulation50mM Tris. pH-8 5mM EDTA.Storage conditionsFive years frozen. One month in solution at room temperature.Tested applicationsSDS-PAGEMarketsource - Marketsource
Specifications
Cat. No.BIRBORB81554-500Protein/peptide nameHCV Genotype-1PurityHCV Core Genotype-1 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-1 protein was purified by proprietary chromatographic technique.Formulation50mM Tris. pH-8 5mM EDTA.Storage conditionsFive years frozen. One month in solution at room temperature.Tested applicationsSDS-PAGEMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



