Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
TAFI
TAFI
TAFI
BIRBORB79690-1
:
Biorbyt
:
TAFI
BIRBORB79690-1
:
Biorbyt
:
Pk:
1 mg
Source:
Species:
Environmentally Preferable:
Storage conditions:
Store at -2 to -8 °C.
Protein/peptide name:
TAFI
Purity:
Greater than 95% pure SDS-PAGE. Protein G Chromatography