Recombinant IL6
Catalog # BIRBORB97777-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Biological activity:EC50:0.5-1.0 ng/ml, calculated by the scattering activityin the MDCK cell assay.
- Protein synonyms:Interleukin BSF 2
- UniProtKB:P14210
- Protein/peptide name:IL6
- Purity:90% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized HGF insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions. Forlong term storage it is recommended to add a carrierprotein (0.1% HSA or BSA).
- Molecular weight:78 kDa
- Formulation:Lyophilized from a solution containing 50mM acetic acid.