Recombinant FGF2
Catalog # BIRBORB97761-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 0.1 ng/ml, corresponding to a specific activity of 2.4x106 Units/mg, measured in a mitogenic assayusing quiescent NR6R-3T3 fibroblasts.
- UniProtKB:P03969
- Protein/peptide name:FGF2
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized FGF insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions.
- Molecular weight:17 kDa
- Formulation:Lyophilized. Recombinant FGF-2 was lyophilized from a1 mg/ml solution containing 1% BSA.